Di-n-butyl sulfide, 98%
Di-n-butyl sulfide, 98%
Di-n-butyl sulfide, 98%
Di-n-butyl sulfide, 98%
Thermo Scientific Chemicals
Di-n-butyl sulfide, 98%
CAS: 544-40-1 | C8H18S | 146.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL07599.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDi-n-butyl sulfide
CAS544-40-1
Health Hazard 1H227-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P312-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P374-P390-P501c
View more
Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-n-butyl sulfide is generally used as an agricultural intermediate, a sulfiding agent, a refinery catalyst, a lubricant additive, a gas odorant and a processing aid in mining applications. It finds application as an internal standard in X-ray fluorescence spectrometry (XRF) measurements of sulfur in oils and other liquid hydrocarbon matrices.
Solubility
Silghtly miscible with water. Miscible with olive oil and almond oil.
Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Buriak, J. M.; Sikder, D. H. From Molecules to Surfaces: Radical-Based Mechanisms of Si-S and Si-Se Bond Formation on Silicon. J. Am. Chem. Soc. 2015, 137 (30), 9730-9738.
  2. Albiter, E.; Alfaro, S.; Valenzuela, M. A. Photosensitized oxidation of 9,10-dimethylanthracene with singlet oxygen by using a safranin O/silica composite under visible light. Photochem. Photobiol. Sci. 2015, 14 (3), 597-602.