3-Methoxypropionitrile, 99%
3-Methoxypropionitrile, 99%
3-Methoxypropionitrile, 99%
3-Methoxypropionitrile, 99%
Thermo Scientific Chemicals
3-Methoxypropionitrile, 99%
CAS: 110-67-8 | C4H7NO | 85.106 g/mol
Have Questions?
Catalog NumberQuantity
ALFL07055.AP500 mL
Catalog number ALFL07055.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material3-Methoxypropionitrile
CAS110-67-8
Health Hazard 1H227-H319
Health Hazard 2GHS H Statement
H319-H335-H227
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
View more
3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxypropionitrile is used as a component in high power lithium ion batteries. It is used as pharmaceutical intermediates. It acts as a good solvent for plastic polymers.
Solubility
Soluble in water. (540 g/L) at 25°C.
Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Hoang Thai Nguyen; Ha Minh Ta; Torben Lund. Thermal thiocyanate ligand substitution kinetics of the solar cell dye N719 by acetonitrile, 3-methoxypropionitrile, and 4-tert-butylpyridine.Solar Energy Materials and Solar Cells.2007,91 1934-1942.
  2. Qing Wang; Shaik M. Zakeeruddin; Ivan Exnar and Michael Grätzel. 3-Methoxypropionitrile-Based Novel Electrolytes for High-Power Li-Ion Batteries with Nanocrystalline Li4Ti5 O 12 Anode.J. Electrochem. Soc.2004,151 A1598-A1603..