2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
2-(2-Aminophenyl)benzimidazole, 98%
Thermo Scientific Chemicals
2-(2-Aminophenyl)benzimidazole, 98%
CAS: 5805-39-0 | C13H11N3 | 209.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFL07034.065 g
Catalog number ALFL07034.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2-(2-Aminophenyl)benzimidazole
CAS5805-39-0
Health Hazard 1H302-H315-H318-H335
Health Hazard 2GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
View more
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(2-Aminophenyl)benzimidazole is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Swadeshmukul Santra; Sneh K.Dogra. Excited-state intramolecular proton transfer in 2-(2'-aminophenyl) benzimidazole. Chemical Physics. 1998, 226,(3), 285-296. 
  2. Pilar Souza; Jose A.Garcia-Vazquez; Jose R.Masaguer. Complexes of divalent nickel and copper with the Schiff base derived from 2-(2-aminophenyl)benzimidazole and benzaldehyde. Transition Metal Chemistry. 1984, 9,(8), 318-321.