Ethyl vinyl ketone, 97%, stab.
Ethyl vinyl ketone, 97%, stab.
Ethyl vinyl ketone, 97%, stab.
Ethyl vinyl ketone, 97%, stab.
Thermo Scientific Chemicals
Ethyl vinyl ketone, 97%, stab.
CAS: 1629-58-9 | C5H8O | 84.118 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL06829.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl vinyl ketone
Name Notestabilized
CAS1629-58-9
Health Hazard 1H225
Health Hazard 2GHS H Statement
H225-H300-H310-H330-H314-H318
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
Ethyl vinyl ketone is used as reagent for annulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl vinyl ketone is used as reagent for annulation.
Solubility
Insoluble in water; soluble in most organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only
General References:
  1. Min Shi; Guang-Ning Ma; Jun Gao. Chiral bifunctional organocatalysts in asymmetric aza-Morita-Baylis-Hillman reactions of ethyl (arylimino) acetates with methyl vinyl ketone and ethyl vinyl ketone. J. Org. Chem. 2007, 72,(25), 9779-9781.
  2. Harold W.Gardner; Marilyn J.Grove; Yangkyo P.Salch. Enzymic pathway to ethyl vinyl ketone and 2-pentenal in soybean preparations. J. Agric. Food Chem. 1996, 44,(3), 882-886.