1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%
Thermo Scientific Chemicals
1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%
CAS: 303180-11-2 | C5H6N2O5 | 174.112 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL06731.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1H-Pyrazole-3,5-dicarboxylic acid monohydrate
CAS303180-11-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c