Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Cyclohexanemethanol, 99%
Thermo Scientific Chemicals
Cyclohexanemethanol, 99%
CAS: 100-49-2 | C7H14O | 114.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL06538.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclohexanemethanol
CAS100-49-2
Health Hazard 1H227
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.
Solubility
Soluble in Ether, Alcohols.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Acid chlorides, Acid anhydrides.
RUO – Research Use Only
General References:
  1. Scott E. Wood. The Thermodynamic Functions of Cyclohexane-Methanol Mixtures.J. Am. Chem. Soc.1946,68 (10), 1963-1966.
  2. C. Houessou; P. Guenoun; R. Gastaud; F. Perrot and D. Beysens. Critical behavior of the binary fluids cyclohexane-methanol, deuterated cyclohexane-methanol and of their isodensity mixture: Application to microgravity simulations and wetting phenomena.Physical Review A.1985,32 1818.