4-Chlorobutyrophenone, 95%, Thermo Scientific™
4-Chlorobutyrophenone, 95%, Thermo Scientific™
4-Chlorobutyrophenone, 95%, Thermo Scientific™
4-Chlorobutyrophenone, 95%, Thermo Scientific™
Thermo Scientific Chemicals
4-Chlorobutyrophenone, 95%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFL06383.065 g
Catalog number ALFL06383.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Chlorobutyrophenone
CAS939-52-6
Melting Point19°C to 20°C
Recommended StorageAmbient temperatures
Density1.137
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.