Octadecyltriethoxysilane, 98%, n-isomer 85% min
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Thermo Scientific Chemicals
Octadecyltriethoxysilane, 98%, n-isomer 85% min
CAS: 7399-00-0 | C24H52O3Si | 416.76 g/mol
Have Questions?
Catalog NumberQuantity
ALFL06366.1850 g
Catalog number ALFL06366.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point165°C to 169°C (0.2mmHg)
Chemical Name or MaterialOctadecyltriethoxysilane
CAS7399-00-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement