Methyl undecanoate, 99%
Methyl undecanoate, 99%
Methyl undecanoate, 99%
Methyl undecanoate, 99%
Thermo Scientific Chemicals
Methyl undecanoate, 99%
CAS: 1731-86-8 | C12H24O2 | 200.322 g/mol
Have Questions?
Catalog NumberQuantity
ALFL05939.065 g
Catalog number ALFL05939.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl undecanoate
CAS1731-86-8
Health Hazard 1H315-H318-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
Melting Point-10°C
View more
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.
Solubility
Soluble in ethanol, and ether. Insoluble in water.
Notes
Store at -20°C. Incompatible with oxidizing agents.
RUO – Research Use Only
Xinhai Yu; Zhenzhong Wen; Hongliang Li; Shan-Tung Tu; Jinyue Yan. Transesterification of Pistacia chinensis oil for biodiesel catalyzed by CaO-CeO2 mixed oxides. Fuel.2011, 90, 1868-1874.