Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Heptadecanoic acid, 98%
Thermo Scientific Chemicals
Heptadecanoic acid, 98%
CAS: 506-12-7 | C17H34O2 | 270.457 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFL05928.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHeptadecanoic acid
CAS506-12-7
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.
Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly). Insoluble in water.
Notes
Store at 4°C. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Chedlia Ben Ahmed; Bechir Ben Rouina; Serhat Sensoy; Mekki Boukhriss. Saline water irrigation effects on fruit development, quality, and phenolic composition of virgin olive oils, cv. Chemlali. Journal of Agricultural and Food Chemistry.2009, 57, (7), 2803-2811.
  2. Gabin Gbabode; Philippe Negrier; Denise Mondieig; Evelyn Moreno; Teresa Calvet; Miquel Àngel Cuevas-Diarte. Polymorphism and solid-state miscibility in the pentadecanoic acid-heptadecanoic acid binary system. Chemistry and Physics of Lipids.2008, 154, (1), 68-77.