3-Chloropropionitrile, 98%
3-Chloropropionitrile, 98%
3-Chloropropionitrile, 98%
3-Chloropropionitrile, 98%
Thermo Scientific Chemicals
3-Chloropropionitrile, 98%
CAS: 542-76-7 | C3H4ClN | 89.52 g/mol
Have Questions?
Catalog NumberQuantity
ALFL05902.22100 g
Catalog number ALFL05902.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloropropionitrile
CAS542-76-7
Health Hazard 1H227-H300-H331
Health Hazard 2GHS H Statement
H300-H227
Fatal if swallowed.
Combustible liquid.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P311-P330-P370+P378q-P501c
View more
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloropropionitrile is used in pharmaceutical and polymer synthesis.
Solubility
Soluble in water. (4.5g/100mL, 25°C). Miscible in Ether, Alcohols, Benzene, Acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong acids, Strong bases, Strong oxidizing agents, Strong reducing agents.
RUO – Research Use Only
General References:
  1. S. Heininger. Notes - Reactions of 3-Chloropropionitrile and Alkyl 3-Chloropropionates with Amines.J. Org. Chem.1957,22 (6), 704-706.
  2. J.R. Durig; G.A. Guirgis; A.S. Drew. Conformational analysis, ab initio calculations, and vibrational assignment of 3-chloropropionitrile.Journal of Molecular Structure.1994,327 55-69.