4-(2-Chloroethyl)morpholine hydrochloride, 99%
4-(2-Chloroethyl)morpholine hydrochloride, 99%
4-(2-Chloroethyl)morpholine hydrochloride, 99%
4-(2-Chloroethyl)morpholine hydrochloride, 99%
Thermo Scientific Chemicals
4-(2-Chloroethyl)morpholine hydrochloride, 99%
CAS: 3647-69-6 | C6H13Cl2NO | 186.076 g/mol
Have Questions?
Catalog NumberQuantity
ALFL05771.36500 g
Catalog number ALFL05771.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(2-Chloroethyl)morpholine hydrochloride
CAS3647-69-6
Health Hazard 1H301-H312-H314-H317-H335
Health Hazard 2GHS H Statement
H301-H314-H318-H312-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
May cause an allergic skin reaction.
Health Hazard 3P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(2-Chloroethyl)morpholine hydrochloride (MOC) is used as intermediate for the synthesis of pharmaceuticals (e.g. floredil, morinamide, nimorazole and pholcodine). It is also used as Agrochemical Intermediates, dyestuff Intermediates.
Solubility
Soluble in water, and methanol.
Notes
Hygroscopic. Do not store together with acids. Store under dry inert gas. Protect from humidity and water. Incompatible materials are acids, moisture.
RUO – Research Use Only