Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
Thermo Scientific Chemicals
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFL05466.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMolecular sieves, 4A
SensitivityHygroscopic
Quantity250 g
Name Note3 to 5mm (0.12 to 0.20 in) beads
CAS70955-01-0
View more
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads are useful in general gas drying applications.