2-(Methylthio)ethanol, 99%
2-(Methylthio)ethanol, 99%
2-(Methylthio)ethanol, 99%
2-(Methylthio)ethanol, 99%
Thermo Scientific Chemicals
2-(Methylthio)ethanol, 99%
CAS: 5271-38-5 | C3H8OS | 92.16 g/mol
Have Questions?
Catalog NumberQuantity
ALFL05442.1425 g
Catalog number ALFL05442.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-(Methylthio)ethanol
CAS5271-38-5
Health Hazard 1H227
Health Hazard 2GHS H Statement
H302-H332-H227
Harmful if swallowed.
Harmful if inhaled.
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Not miscible in water.
Notes
Do not store together with acids. Store away from Oxidizing agents.
RUO – Research Use Only
General References:
  1. Michael L. McKee. Comparison of Gas-Phase and Solution-Phase Reactions of Dimethyl Sulfide and 2-(Methylthio)ethanol with Hydroxyl Radical.J. Phys. Chem. A.2003,107 (35), 6819-6827.
  2. Reagent for carboxyl group protection in peptide synthesis. Cleavage is by alkylation (MeI), followed by mild alkaline hydrolysis: J. Chem. Soc. (C), 807 (1966); Chem. Ber., 109, 3693 (1976). See Appendix 6.