2-Mercaptothiazoline, 98+%
2-Mercaptothiazoline, 98+%
2-Mercaptothiazoline, 98+%
2-Mercaptothiazoline, 98+%
Thermo Scientific Chemicals
2-Mercaptothiazoline, 98+%
CAS: 96-53-7 | C3H5NS2 | 119.2 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04924.22100 g
Catalog number ALFL04924.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Mercaptothiazoline
CAS96-53-7
Health Hazard 1H301
Health Hazard 2GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3P264b-P270-P301+P310-P330-P501c
View more
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.
Solubility
Soluble in chloroform, methanol. Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Lei Tao et. al. Synthesis and bioactivity of poly(HPMA)-lysozyme conjugates: the use of novel thiazolidine-2-thione coupling chemistry. Organic & Biomolecular Chemistry. 2009, 7,(17), 3481-3485 .
  2. Newton L Dias Filho et. al. Preparation of a clay-modified carbon paste electrode based on 2-thiazoline-2-thiol-hexadecylammonium sorption for the sensitive determination of mercury. Analytical Sciences. 2005, 21,(11), 1309-1316 .
  3. Used in the chain-lengthening of alkyl halides by trans-iodopropenylenation: Org. Synth. Coll., 6, 704 (1988):
  4. Reaction with ɑ-bromo esters gives the S-alkoxycarbonylmethyl derivatives, which can be C-alkylated by alkyl halides, using NaH as base. Sulfur-free products can be obtained by reduction (Zn/AcOH) to give the substituted acetic acid, or by reaction with iodidomethane to give the corresponding ɑ-iodo ester: Tetrahedron Lett., 2677 (1977).