2,3-Hexanedione, 94%
2,3-Hexanedione, 94%
2,3-Hexanedione, 94%
2,3-Hexanedione, 94%
Thermo Scientific Chemicals
2,3-Hexanedione, 94%
CAS: 3848-24-6 | C6H10O2 | 114.14 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04669.1425 g
Catalog number ALFL04669.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2,3-Hexanedione
CAS3848-24-6
Health Hazard 1H226-H315-H319-H335
Health Hazard 2GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
View more
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Hexanedione is used to evaluate the influence of xanthan concentration on the release of aroma compounds in xanthan-thickened food model systems. It reacts with ethylenediamine to yield macrocyclic tetradentate 12-membered nitrogen donor (N4) ligand.
Solubility
Partly miscible in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. James W. Hall.; Wayne E. Marsh.; Robert R. Weller.; William E. Hatfield. Exchange coupling in the alternating-chain compounds catena-di-.mu.-chloro-bis(4-methylpyridine)copper(II), catena-di-.mu.-bromobis(N-methylimidazole)copper(II), catena-[hexanedione)bis(thiosemicarbazonato)]copper(II), and catena-[octanedione bis (thiosemicarbazonato)]copper(II). Inorg. Chem. 1991, 20 (4), 1033-1037.
  2. Robert Chenevert.; Sonia Thiboutot. Baker’s Yeast Reduction of 1,2-Diketones. Preparation of Pure (S)-(-)-2-Hydroxy-1-phenyl-1-propanone. Chemistry Letters. 1988, 17 (7), 1191-1192.