1,1,2,2-Tetrachloroethane, 98+%
1,1,2,2-Tetrachloroethane, 98+%
1,1,2,2-Tetrachloroethane, 98+%
1,1,2,2-Tetrachloroethane, 98+%
Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane, 98+%
CAS: 79-34-5 | C2H2Cl4 | 167.838 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04489.1425 g
Catalog number ALFL04489.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,1,2,2-Tetrachloroethane
CAS79-34-5
Health Hazard 1H310+H330-H351
Health Hazard 2GHS H Statement
H310-H330-H341-H351
Fatal in contact with skin.
Fatal if inhaled.
Suspected of causing genetic defects.
Suspected of causing cancer.
Health Hazard 3P201-P202-P260-P262-P264b-P270-P271-P281-P284-P302+P350-P304+P340-P308+P313-P310-P361-P363-P501c
View more
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.
Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.
Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
RUO – Research Use Only
General References:
  1. Tan, C.; Xiang, H.; He, Q.; Yang, C. A One-Pot Copper(II)-Catalyzed Tandem Synthesis of 2-Substituted Pyrrolo[1,2-b]pyridazin-4(1H)-ones. Eur. J. Org. Chem. 2015, 2015 (17), 3656-3660.
  2. Chen, W.; Li, Y.; Zhu, D.; Zheng, S.; Chen, W. Dehydrochlorination of activated carbon-bound 1,1,2,2-tetrachloroethane: Implications for carbonaceous material-based soil/sediment remediation. Carbon 2014, 78, 578-588.