n-Octyltriethoxysilane, 95%
n-Octyltriethoxysilane, 95%
n-Octyltriethoxysilane, 95%
n-Octyltriethoxysilane, 95%
Thermo Scientific Chemicals
n-Octyltriethoxysilane, 95%
CAS: 2943-75-1 | C14H32O3Si | 276.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1000 g
Catalog number ALFL04407.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point265°C
Chemical Name or Materialn-Octyltriethoxysilane
Melting Point<-40°C
CAS2943-75-1
Health Hazard 1H315-H319