1-Chlorooctane, 99%
1-Chlorooctane, 99%
1-Chlorooctane, 99%
Thermo Scientific Chemicals
1-Chlorooctane, 99%
CAS: 111-85-3 | C8H17Cl | 148.674 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFL04358.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Chlorooctane
CAS111-85-3
Health Hazard 1H227
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c