1,2,3-Trichloropropane, 98+%
1,2,3-Trichloropropane, 98+%
1,2,3-Trichloropropane, 98+%
1,2,3-Trichloropropane, 98+%
Thermo Scientific Chemicals
1,2,3-Trichloropropane, 98+%
CAS: 96-18-4 | C3H5Cl3 | 147.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04312.30250 g
Catalog number ALFL04312.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,2,3-Trichloropropane
CAS96-18-4
Health Hazard 1H227-H302+H312-H350-H360F
Health Hazard 2GHS H Statement
H350-H360-H302-H312-H332-H227
May cause cancer.
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Combustible liquid.
Health Hazard 3P201-P202-P210-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P370+P378q-P501c
View more
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,3-Trichloropropane is used to study its reaction with OH regarding it spotential atmospheric relevance.
Solubility
Slightly soluble in water (2 g/L at 25°C).
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
RUO – Research Use Only
General References:
  1. Alexandra J. Salter-Blanc.; Paul G. Tratnyek. Effects of Solution Chemistry on the Dechlorination of 1,2,3-Trichloropropane by Zero-Valent Zinc. Environ. Sci. Technol. 2011, 45 (9), 4073-4079.
  2. J. Yan.; B. A. Rash.; F. A. Rainey.; W. M. Moe. Isolation of novel bacteria within the Chloroflexi capable of reductive dechlorination of 1,2,3-trichloropropane. Environmental Microbiology. 2009, 11 (4), 833-8431.