Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
Thermo Scientific Chemicals
Acrylic acid, 97%, stab. with ca 200ppm 4-methoxyphenol
CAS: 79-10-7 | C3H4O2 | 72.063 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04280.36500 g
Catalog number ALFL04280.36
Price (MYR)
443.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
443.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcrylic acid
Name Notestabilized with ≈200ppm 4-methoxyphenol
CAS79-10-7
Health Hazard 1H226-H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
View more
Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrylic acid acts as a precursor in the production of 3-hydroxypropionic acid. It is used in the preparation of water-absorbent resins. It reacts with alcohols to prepare the corresponding esters. Its esters are used as raw materials for synthetic resins, rubbers, coating adhesives, water-based paints, floor polishes and adhesives. It is also used to prepare forming homopolymers or copolymers by reacting with other monomers like acrylamides, acrylonitrile, vinyl, styrene, and butadiene.
Solubility
Miscible with water.
Notes
Hygroscopic. Light and air sensitive. Incompatible with strong oxidizing agents, strong bases, oxygen, polymerizing initiators, amines and peroxides.
RUO – Research Use Only
General References:
  1. Börjesson, M.; Richardson, G.; Westman, G. UV Radiation of Cellulose Fibers and Acrylic Acid Modified Cellulose Fibers for Improved Stiffness in Paper. Bioresources 2015, 10 (2), 3056-3069.
  2. Yamaguchi, H.; Nakanishi, S.; Iba, H.; Itoh, T. Amorphous polymeric anode materials from poly(acrylic acid) and tin(II) oxide for lithium ion batteries. J. Power Sources 2015, 275, 1-5.