Propionic acid, 99%
Propionic acid, 99%
Propionic acid, 99%
Thermo Scientific Chemicals
Propionic acid, 99%
CAS: 79-09-4 | C3H6O2 | 74.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1000 g
2500 g
Catalog number ALFL04210.22
Price (MYR)
275.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
275.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropionic acid
CAS79-09-4
Health Hazard 1H226-H290-H314-H335
Health Hazard 2GHS H Statement
H314-H226
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
View more
Propionic acid is used as a preservative for animal feed and food for human consumption. It acts as an intermediate in the production of polymers like cellulose-acetate-propionate, artificial flavorings, agrochemicals and in bleaching agents. In addition, it is used in the synthesis of morphans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propionic acid is used as a preservative for animal feed and food for human consumption. It acts as an intermediate in the production of polymers like cellulose-acetate-propionate, artificial flavorings, agrochemicals and in bleaching agents. In addition, it is used in the synthesis of morphans.
Solubility
Miscible with water, alcohol, diethyl ether and chloroform.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Wang, Z.; Jin, Y.; Yang, S. T. High cell density propionic acid fermentation with an acid tolerant strain of Propionibacterium acidipropionici. Biotechnol. Bioeng. 2015, 112 (3), 502-511.
  2. Zhang, A.; Sun, J.; Wang, Z.; Yang, S. T.; Zhou, H. Effects of carbon dioxide on cell growth and propionic acid production from glycerol and glucose by Propionibacterium acidipropionici. Bioresour. Technol. 2015, 175, 3716-3722.