Sulfamerazine, 98+%
Sulfamerazine, 98+%
Sulfamerazine, 98+%
Thermo Scientific Chemicals
Sulfamerazine, 98+%
CAS: 127-79-7 | C11H12N4O2S | 264.303 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL04194.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfamerazine
CAS127-79-7
Melting Point234°C to 238°C
Recommended StorageAmbient temperatures
RTECS NumberWP0750000