1-Octadecanethiol, 96%
1-Octadecanethiol, 96%
1-Octadecanethiol, 96%
1-Octadecanethiol, 96%
Thermo Scientific Chemicals
1-Octadecanethiol, 96%
CAS: 2885-00-9 | C18H38S | 286.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL04123.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Octadecanethiol
CAS2885-00-9
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
View more
1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octadecanethiol (ODT) is used to deposit thiolated molecules onto the surface of gold substrates, such as SAMs on gold quartz crystal resonators, on the tip of atomic force microscope probes, capacitive sensors.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only
General References:
  1. Tao Zhang; Zengguang Cheng; Yibing Wang, Zhongjun Li; Chenxuan Wang; Yibao Li; and Ying Fang. Self-Assembled 1-Octadecanethiol Monolayers on Graphene for Mercury Detection.Nano Lett.1994, 10 (11), 4738-4741.
  2. Ken Mullen; Keith Carron. Adsorption of Chlorinated Ethylenes at 1-Octadecanethiol-Modified Silver Surfaces.Anal. Chem.1994, 66 (4), 478-483.