2,3-Dimercaptopropanol, 97%
2,3-Dimercaptopropanol, 97%
2,3-Dimercaptopropanol, 97%
2,3-Dimercaptopropanol, 97%
Thermo Scientific Chemicals
2,3-Dimercaptopropanol, 97%
CAS: 59-52-9 | C3H8OS2 | 124.216 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03953.031 g
Catalog number ALFL03953.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2,3-Dimercaptopropanol
CAS59-52-9
Health Hazard 1H301-H315-H317-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H317
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
View more
British Anti-Lewisite. A chelator that inhibits sugar nucleotide degradation.2,3-Dimercaptopropanol is used as a chelating agent involved in the treatment of arsenic, antimony, lead, gold and mercury poisoning. It is also used as an antidote to the warfare agent Lewisite. Further, it is used for the treatment of Wilson's disease. In addition to this, it is used as an adjunct in the treatment of the acute encephalopathy of lead toxicity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Miscible with water, ether, ethanol, vegetable oils and acetone. Slightly miscible with chloroform.
Notes
Air sensitive. Store in a cool place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
RUO – Research Use Only
General References:
  1. Chio, C. M.; Cheng, K. W.; Bishop, A. C. Direct Chemical Activation of a Rationally Engineered Signaling Enzyme. ChemBioChem 2015, 16 (12), 1735-1739.
  2. Andries, C.; Manea, M.; Drochioiu, G.; Gradinaru, R. New insights into coenzyme A interaction with mercury ions provided by mass spectrometric and circular dichroism spectroscopic approaches. Eur. J. Mass Spectrom. 2015, 21 (2), 97-102.