1-Pentadecanol, 99%
1-Pentadecanol, 99%
1-Pentadecanol, 99%
Thermo Scientific Chemicals
1-Pentadecanol, 99%
CAS: 629-76-5 | C15H32O | 228.42 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03673.065 g
Catalog number ALFL03673.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Pentadecanol
CAS629-76-5
Melting Point45°C to 46°C
Recommended StorageAmbient temperatures
EINECS Number211-107-9
View more
1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.
Solubility
Insoluble in water.
Notes
Incompatible materials are Strong oxidizing a
RUO – Research Use Only
General References:
  1. K. Venkatesan; K.V. Srinivasan. A novel stereoselective synthesis of pachastrissamine (jaspine B) starting from 1-pentadecanol.Tetrahedron: Asymmetry.2008, 19 209-215.
  2. J. Cees van Miltenburg and Gerrit. J. K. van den Berg. Heat Capacities and Derived Thermodynamic Functions of 1-Dodecanol and 1-Tridecanol between 10 K and 370 K and Heat Capacities of 1-Pentadecanol and 1-Heptadecanol between 300 K and 380 K and Correlations for the Heat Capacity and the Entropy of Liquid n-Alcohols.J. Chem. Eng. Data.2003, 48 (1), 36-43.