Thioacetic acid, 97%
Thioacetic acid, 97%
Thioacetic acid, 97%
Thioacetic acid, 97%
Thermo Scientific Chemicals
Thioacetic acid, 97%
CAS: 507-09-5 | C2H4OS | 76.11 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03305.1425 g
Catalog number ALFL03305.14
Price (MYR)
332.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
332.00
EA
Add to cart
Specifications
Chemical Name or MaterialThioacetic acid
CAS507-09-5
Health Hazard 1H225-H301-H317-H318-H335
Health Hazard 2GHS H Statement
H225-H314-H318-H302-H332-H317
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Health Hazard 3P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P333+P313-P363-P370+P378q-P501c
View more
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).
Solubility
Soluble in water. (27g/L) at 16°C.
Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents, metals.
RUO – Research Use Only
General References:
  1. Wen Yang; Da-Ming Du. Squaramide-catalysed enantio- and diastereoselective sulfa-Michael addition of thioacetic acid to α,β-disubstituted nitroalkenes.Organic & Biomolecular Chemistry.2012, 10 (34), 6876-6884.
  2. Libo Hu; Hui Zhu; Da-Ming Du; Jiaxi Xu. Efficient synthesis of taurine and structurally diverse substituted taurines from aziridines.Journal of Organic Chemistry.2007, 72 (12), 4543-4546.
  3. Used in several different ways for the introduction of the thiol group: Radical addition to unactivated alkenes: J. Chem. Soc., 2123 (1951). Conjugate (Michael) addition to activated double bonds: J. Am. Chem. Soc., 77, 5144 (1955); 81, 1224 (1959). Nucleophilic displacement of halide, tosylate or acetate: J. Am. Chem. Soc., 73, 2659 (1951); Carbohydr. Res., 17, 457 (1971). Alcohols can be converted to thiol esters under Mitsunobu conditions, followed by LAH reduction to the thiol: Tetrahedron Lett., 3119 (1981).