Solketal, 97%
Solketal, 97%
Solketal, 97%
Thermo Scientific Chemicals
Solketal, 97%
CAS: 100-79-8 | C6H12O3 | 132.159 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02814.36500 g
Catalog number ALFL02814.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSolketal
CAS100-79-8
Health Hazard 1H227-H319
Health Hazard 3P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
Melting Point-27°C
View more
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Solketal is useful for synthesis of mono-, di- and triglycerides. It is used as the starting reagent for synthesis of tulipaline derivatives. It acts as a fuel additive in gasoline. It is an inhibitor of Methyl ethyl ketone .
Solubility
Soluble in water (fully), alcohol, acetals, esters, and ethers.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are acids, Strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Claudio J. A. Mota; Carolina X. A. da Silva;Nilton Rosenbach; Jair Costa and Flávia da Silva. Glycerin Derivatives as Fuel Additives: The Addition of Glycerol/Acetone Ketal (Solketal) in Gasolines.Energy Fuels.2010, 24 (4), 2733-2736.
  2. Jan-Willem Hofman; Femke van Zeeland; Selcan Turker; Herre Talsma; Saskia A. G. Lambrechts; Dmitri V. Sakharov; Wim E. Hennink and Cornelus F. van Nostrum. Peripheral and Axial Substitution of Phthalocyanines with Solketal Groups: Synthesis and In Vitro Evaluation for Photodynamic Therapy.J. Med. Chem.2007, 50 (7), 1485-1494.