Diphenyl-n-propylphosphine, 97%
Diphenyl-n-propylphosphine, 97%
Diphenyl-n-propylphosphine, 97%
Diphenyl-n-propylphosphine, 97%
Thermo Scientific Chemicals
Diphenyl-n-propylphosphine, 97%
CAS: 7650-84-2 | C15H17P | 228.28 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02402.065 g
Catalog number ALFL02402.06
Price (MYR)
706.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
706.00
EA
Add to cart
Specifications
Chemical Name or MaterialDiphenyl-n-propylphosphine
CAS7650-84-2
Health Hazard 1H302-H315-H319
Health Hazard 2GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c