2-Methoxybenzylamine, 98+%
2-Methoxybenzylamine, 98+%
2-Methoxybenzylamine, 98+%
2-Methoxybenzylamine, 98+%
Thermo Scientific Chemicals
2-Methoxybenzylamine, 98+%
CAS: 6850-57-3 | C8H11NO | 137.182 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02273.1425 g
Catalog number ALFL02273.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Methoxybenzylamine
CAS6850-57-3
Health Hazard 1H314
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
RUO – Research Use Only
General References:
  1. Kouichi Takahashi.; Hiroto Nakano.; Reiko Fujita. A concise and practical catalytic asymmetric synthesis of (-)-CP-99,994 and (-)-L-733,061. Tetrahedron Letters. 2005, 46 (51), 8927-8930.
  2. Kazuya Yamaguchi Dr.; Noritaka Mizuno Prof. Dr. Efficient Heterogeneous Aerobic Oxidation of Amines by a Supported Ruthenium Catalyst. Angewandte Chemie International Edition. 2003, 42(13), 1480-1483.