4-Ethyltoluene, 97%
4-Ethyltoluene, 97%
4-Ethyltoluene, 97%
Thermo Scientific Chemicals
4-Ethyltoluene, 97%
CAS: 622-96-8 | C9H12 | 120.195 g/mol
Have Questions?
Catalog NumberQuantity
ALFL02120.065 g
Catalog number ALFL02120.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Ethyltoluene
CAS622-96-8
Health Hazard 1H226-H304
Health Hazard 2GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
View more
4-Ethyltoluene is used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethyltoluene is used as intermediates.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only
General References:
  1. Consuelo Pizarro; Octavio Suárez-Iglesias; Ignacio Medina and Julio L. Bueno. Binary Diffusion Coefficients of 2-Ethyltoluene, 3-Ethyltoluene, and 4-Ethyltoluene in Supercritical Carbon Dioxide.J. Chem. Eng. Data.2009, 54 (5), 1467-1471.
  2. Tomas T; Swiercz R; Wiaderna D. Changes in function of rabbit cerebral cortex after intraperitoneal injections of organic solvents. II. Pseudocumene and 4-ethyltoluene.Medycyna Pracy.1997, 48 (3), 307-315.