1-Naphthalenemethanol, 98+%
1-Naphthalenemethanol, 98+%
1-Naphthalenemethanol, 98+%
1-Naphthalenemethanol, 98+%
Thermo Scientific Chemicals
1-Naphthalenemethanol, 98+%
CAS: 4780-79-4 | C11H10O | 158.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFL01347.1850 g
Catalog number ALFL01347.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Naphthalenemethanol
CAS4780-79-4
Melting Point58°C to 63°C
Recommended StorageAmbient temperatures
RTECS NumberQJ8880000