Check Order Status
Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure
Element Search
Structure Search
Thermo Scientific Chemicals
Tris, 99.8 to 100.1% (Dry Basis), Molecular Biology Grade, Ultrapure
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
ALFJ75825.36
500 g
Plastic bottle
Catalog number
ALFJ75825.36
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Tris
CAS
77-86-1
Melting Point
171°C to 172°C
Boiling Point
219°C to 220°C (10 mmHg)
Recommended Storage
Ambient temperatures