Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade
Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade
Thermo Scientific Chemicals
Linear polyacrylamide solution for DNA/RNA precipitation, 5mg/ml, Molecular Biology Grade
Have Questions?
Catalog NumberQuantity
ALFJ67830.XF1 mL
Catalog number ALFJ67830.XF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 mL
Request bulk or custom format
Specifications
Recommended StorageStore at -20°C
ColorClear, colorless
Product TypeLinear polyacrylamide solution
Quantity1 mL
SynonymLPA solution for DNA/RNA precipitation, Molecular Biology Grade;
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C