Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold nanoparticles, 250nm, supplied in 0.1mM PBS, reactant-free, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ67457.AC25 mL
Catalog number ALFJ67457.AC
Price (MYR)
1,035.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
1,035.00
EA
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorBrown to Pink
Name Notesupplied in 0.1mM PBS, reactant-free
CAS7440-57-5
Storage Note 12° to 8°C
View more
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications