Boric acid, Molecular Biology Grade, 99.5% min
Boric acid, Molecular Biology Grade, 99.5% min
Boric acid, Molecular Biology Grade, 99.5% min
Boric acid, Molecular Biology Grade, 99.5% min
Thermo Scientific Chemicals
Boric acid, Molecular Biology Grade, 99.5% min
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ67419.A11 kg
Catalog number ALFJ67419.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialBoric acid
pH3.8 to 4.8
ColorWhite
CAS10043-35-3
Melting Point∼185°C (decomposition)
View more
Used to promote amidations of carboxylic acids and amines in catalytic amounts.