Gold nanoparticles, 30nm, supplied in 0.1mM PBS, 95%, OD1, 526nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 30nm, supplied in 0.1mM PBS, 95%, OD1, 526nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 30nm, supplied in 0.1mM PBS, 95%, OD1, 526nm absorption, Thermo Scientific Chemicals
Gold nanoparticles, 30nm, supplied in 0.1mM PBS, 95%, OD1, 526nm absorption, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold nanoparticles, 30nm, supplied in 0.1mM PBS, 95%, OD1, 526nm absorption, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFJ67368.AC25 mL
Catalog number ALFJ67368.AC
Price (MYR)
727.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
727.00
EA
Add to cart
Specifications
Chemical Name or MaterialGold nanoparticles
ColorRed
Name Notesupplied in 0.1mM PBS, 95%, OD: 1, 526nm absorption
CAS7440-57-5
Storage Note 12° to 8°C
View more
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
RUO – Research Use Only