Cycloheximide, ultrapure, 95%
Cycloheximide, ultrapure, 95%
Cycloheximide, ultrapure, 95%
Cycloheximide, ultrapure, 95%
Thermo Scientific Chemicals
Cycloheximide, ultrapure, 95%
CAS: 66-81-9 | C15H23NO4 | 281.35 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66901.031 g
Catalog number ALFJ66901.03
Price (MYR)
519.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
519.00
EA
Add to cart
Specifications
Chemical Name or MaterialCycloheximide
TypeCycloheximide
Name NoteUltrapure
Physical FormPowder
CAS66-81-9
View more
A protein synthesis inhibitor in eukaryotic cells