Amido Black 10B, Electrophoresis Reagent
Amido Black 10B, Electrophoresis Reagent
Amido Black 10B, Electrophoresis Reagent
Amido Black 10B, Electrophoresis Reagent
Thermo Scientific Chemicals
Amido Black 10B, Electrophoresis Reagent
CAS: 1064-48-8 | C22H14N6Na2O9S2 | 616.487 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALFJ66798.1425 gPlastic bottle
Catalog number ALFJ66798.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmido Black 10B
CAS1064-48-8
Health Hazard 1H315-H317-H319-H335-H372-H500
Health Hazard 3P261-P264b-P270-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P314-P332+P313-P362-P501c
Recommended StorageAmbient temperatures
View more
Used to stain proteins in polyacrylamide gels.