2-Mercaptoethanol, Electrophoresis Grade, 98+%
2-Mercaptoethanol, Electrophoresis Grade, 98+%
2-Mercaptoethanol, Electrophoresis Grade, 98+%
2-Mercaptoethanol, Electrophoresis Grade, 98+%
Thermo Scientific Chemicals
2-Mercaptoethanol, Electrophoresis Grade, 98+%
CAS: 60-24-2 | C2H6OS | 78.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFJ66742.30
Price (MYR)
315.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
315.00
EA
Add to cart
Specifications
Chemical Name or Material2-Mercaptoethanol
CAS60-24-2
Health Hazard 1H227-H301+H331-H310-H315-H317-H318-H373
Health Hazard 2GHS H Statement
H301-H310-H318-H227-H315-H317-H335
Toxic if swallowed.
Fatal in contact with skin.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P210-P235-P260-P262-P264b-P270-P271-P272-P280-P301+P310-P302+P350-P304+P340-P305+P351+P338-P310-P311-P314-P330-P333+P313-P361-P363-P370+P378q-P501c
View more
β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.
Solubility
Fully miscible in water. Soluble in alcohol, ether, benzene, and other organic solvents.
Notes
Handle under dry protective gas. Store in cool condition. Incompatible with air, moisture, oxidizing agents.
RUO – Research Use Only
General References:
  1. Margaret L. Heidrick..; Linda C. Hendricks.; David E. Cook. Effect of dietary 2-mercaptoethanol on the life span, immune system, tumor incidence and lipid peroxidation damage in spleen lymphocytes of aging BC3F1 mice. Mechanisms of Ageing and Development. 1984, 27 (3),341-358 .
  2. B. Tasheva.; G. Dessev. Artifacts in sodium dodecyl sulfate-polyacrylamide gel electrophoresis due to 2-mercaptoethanol.Anal. Biochem. 1983, 129 (1),98-102 .