Colistin sodium methanesulfonate
Colistin sodium methanesulfonate
Colistin sodium methanesulfonate
Colistin sodium methanesulfonate
Thermo Scientific Chemicals
Colistin sodium methanesulfonate
CAS: 8068-28-8 | C58H105N16Na5O28S5 | 1749.811 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66698.031 g
Catalog number ALFJ66698.03
Price (MYR)
1,205.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
1,205.00
EA
Add to cart
Specifications
Chemical Name or MaterialColistin sodium methanesulfonate
TypeColistin Sodium Methanesulfonate
Physical FormPowder
CAS8068-28-8
Storage Note 12° to 8°C
View more
Colistin sodium methanesulfate is used to permeabilize bacterial cell membranes and to study mannose-resistant haemagglutination. It is used to study effective treatments of P. aeruginosa infections in cystic fibrosis patients and to study the pharmacokinetics and BAL concentration of colistin in critically ill patients
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Freely soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
General References:
  1. Kimberly J. Griffitt.; D. Jay Grimes. A novel agar formulation for isolation and direct enumeration of Vibrio vulnificus from oyster tissue. Journal of Microbiological Methods. 2013, 94, (2), 98-102.
  2. Masamichi Fukuda.; Kazuyuki Sasaki. General Purpose Antimicrobial Ophthalmic Solutions Evaluated Using New Pharmacokinetic Parameter of Maximum Drug Concentration in Aqueous. Japanese Journal of Ophthalmology. 2002, 46, (4), 384-390.