Ampicillin trihydrate
Ampicillin trihydrate
Ampicillin trihydrate
Ampicillin trihydrate
Thermo Scientific Chemicals
Ampicillin trihydrate
Commonly used to select for ampicillin resistance in mutated and transformed cells
Have Questions?
Catalog NumberQuantity
ALFJ66514.1425 g
Catalog number ALFJ66514.14
Price (MYR)
642.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
642.00
EA
Add to cart
Chemical Identifiers
CAS7177-48-2
Specifications
CAS7177-48-2
Health Hazard 1H317-H334-H335
Health Hazard 2GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Chemical Name or MaterialAmpicillin trihydrate
Health Hazard 3P261-P271-P272-P280g-P285-P302+P352-P304+P340-P333+P313-P342+P311-P363-P501c
View more
Commonly used to select for ampicillin resistance in mutated and transformed cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
Keep cold