Meropenem trihydrate, 98-101%
Meropenem trihydrate, 98-101%
Meropenem trihydrate, 98-101%
Meropenem trihydrate, 98-101%
Thermo Scientific Chemicals
Meropenem trihydrate, 98-101%
CAS: 119478-56-7 | C17H25N3O5S | 383.46 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66446.MF50 mg
Catalog number ALFJ66446.MF
Price (MYR)
1,462.00
EA
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (MYR)
1,462.00
EA
Add to cart
Specifications
Chemical Name or MaterialMeropenem trihydrate
TypeMeropenem Trihydrate
Physical FormPowder
CAS119478-56-7
Health Hazard 1H315-H319-H335
View more
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.
Solubility
Soluble in aqueous solution at approximately 5mg/ml
Notes
RUO – Research Use Only
General References:
  1. Hiroshi Yamamoto.; Fumio Yamamoto.; Kazuyuki Ishibashi.; Mamika Motokawa. In situ replacement with equine pericardial roll grafts for ruptured infected aneurysms of the abdominal aorta. Journal of Vascular Surgery. 2009, 49, (4), 1041-1045.
  2. Hiromichi Yamaguchi.; Yuko Komase.; Mizuki Ikehara.; Takahito Yamamoto.; Toshihito Shinagawa. Disseminated cryptococcal infection with eosinophilia in a healthy person. Journal of Infection and Chemotherapy. 2008, 14, (4), 319-324.