Flumequine, 98%
Flumequine, 98%
Flumequine, 98%
Thermo Scientific Chemicals
Flumequine, 98%
Inhibits topoisomerases in gram negative bacteria | CAS: 42835-25-6 | C14H12FNO3 | 261.252 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66332.031 g
Catalog number ALFJ66332.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFlumequine
TypeFlumequine
Physical FormCrystalline Powder
CAS42835-25-6
Melting Point253°C to 255°C
View more
Inhibits topoisomerases in Gram-negative bacteria.