N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Thermo Scientific Chemicals
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
For protein solubilization | CAS: 14933-08-5 | C17H37NO3S | 335.547 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66265.065 g
Catalog number ALFJ66265.06
Price (MYR)
663.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
663.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
CAS14933-08-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
For protein solubilization