Piperacillin, 95%
Piperacillin, 95%
Piperacillin, 95%
Thermo Scientific Chemicals
Piperacillin, 95%
CAS: 61477-96-1 | C23H26N5NaO7S | 539.54 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66143.ME500 mg
Catalog number ALFJ66143.ME
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialPiperacillin
TypePiperacillin
Physical FormPowder
CAS61477-96-1
Health Hazard 1H317-H334
View more
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.
Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only
General References:
  1. Gerald P. Bodey; and Barbara Le Blanc. Piperacillin: In Vitro Evaluation. Antimicrob. Agents Chemother., 1978,14(1), 78-87.
  2. N.A Kuck: G.S. Redin. In vitro and in vivo activity of piperacillin, a new broad-spectrum semisynthetic penicillin. The Journal of Antibiotics, 1978,31(11), 1175-1182.