Brij∣r L23, Electrophoresis Reagent
Brij∣r L23, Electrophoresis Reagent
Brij∣r L23, Electrophoresis Reagent
Brij∣r L23, Electrophoresis Reagent
Brij∣r L23, Electrophoresis Reagent
Thermo Scientific Chemicals
Brij∣r L23, Electrophoresis Reagent
CAS: 9002-92-0
Have Questions?
Catalog NumberQuantity
ALFJ66096.30250 g
Catalog number ALFJ66096.30
Price (MYR)
273.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
273.00
EA
Add to cart
Chemical Identifiers
CAS9002-92-0
Specifications
Chemical Name or MaterialBrij™ L23
ColorWhite
Name NoteElectrophoresis Reagent
CAS9002-92-0
Health Hazard 1Warning