Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Toluidine Blue O, Certified, 65% min dye content
Thermo Scientific Chemicals
Toluidine Blue O, Certified, 65% min dye content
CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
25 g
Catalog number ALFJ66015.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialToluidine Blue O
CAS92-31-9
Recommended StorageAmbient temperatures
RTECS NumberSP5670000
EINECS Number202-146-2