Cesium chloride, ultrapure, 99.9% (metals basis)
Cesium chloride, ultrapure, 99.9% (metals basis)
Cesium chloride, ultrapure, 99.9% (metals basis)
Cesium chloride, ultrapure, 99.9% (metals basis)
Thermo Scientific Chemicals
Cesium chloride, ultrapure, 99.9% (metals basis)
CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ65950.1425 g
Catalog number ALFJ65950.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Point1303°C
Chemical Name or MaterialCesium chloride
Melting Point646°C
CAS7647-17-8