Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
Thermo Scientific Chemicals
Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%
CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ65585.36500 g
Catalog number ALFJ65585.36
Price (MYR)
578.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
578.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylenediaminetetraacetic acid
CAS60-00-4
Health Hazard 1Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after repeated exposure (category 2)
Health Hazard 2Causes serious eye irritation, Harmful if inhaled, May cause damage to organs through prolonged or repeated exposure if inhaled
Melting Point∽245°C (decomposition)